Learn More
Biomatik Corporation Recombinant Angiopoietin Like Protein 4 (ANGPTL4), 100UG
Supplier:  Biomatik Corporation RPU40272
Encompass_Preferred
Recombinant Rat Angiopoietin Like Protein 4 (ANGPTL4) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Angiopoietin Like Protein 4 (ANGPTL4). Target Synonyms: ARP4; FIAF; HFARP; NL2; PGAR; Pp1158; Fasting-Induced Adipose Factor; Hepatic Angiopoietin-Related Protein; Hepatic Fibrinogen/Angiopoietin-Related Protein. Accession Number: Q6TMA8. Gene ID: 362850. Expression Region: Arg182~Leu394. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5466