Learn More
Biomatik Corporation Recombinant Annexin A5 (ANXA5), 100UG
Supplier:  Biomatik Corporation RPU40315
Encompass_Preferred
Recombinant Human Annexin V (ANXA5) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Annexin V (ANXA5). Target Synonyms: ANX5; ENX2; PP4; PAP-I; CBP-I; Anchorin CII; Calphobindin I; Endonexin II; Lipocortin V; Placental anticoagulant protein 4; Thromboplastin inhibitor; Vascular anticoagulant-alpha. Accession Number: P08758. Gene ID: 308. Expression Region: Met1~Asp320. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5470