Learn More
Biomatik Corporation Recombinant Anti-Mullerian Hormone (AMH), 100UG
Supplier:  Biomatik Corporation RPU40323
Encompass_Preferred
Recombinant Rat Anti-Mullerian Hormone (AMH) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Anti-Mullerian Hormone (AMH). Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Accession Number: P49000. Gene ID: 25378. Expression Region: Lys447~Arg553. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5472