Learn More
Biomatik Corporation Recombinant Apolipoprotein A1 (APOA1), 100UG
Supplier:  Biomatik Corporation RPU40330
Encompass_Preferred
Recombinant Human Apolipoprotein A1 (APOA1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Apolipoprotein A1 (APOA1). Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Accession Number: P02647. Gene ID: 335. Expression Region: Gln122~Gln267. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5473