Learn More
Biomatik Corporation Recombinant Apolipoprotein C2 (APOC2), 100UG
Supplier:  Biomatik Corporation RPU40352
Encompass_Preferred
Recombinant Mouse Apolipoprotein C2 (APOC2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Apolipoprotein C2 (APOC2). Target Synonyms: APC2; Apo-C2; ProapoC-II; Apolipoprotein C-II; Proapolipoprotein C-II. Accession Number: Q05020. Gene ID: 11813. Expression Region: Ile13~Glu97. Tag Info: N-terminal His and GST Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5475