Learn More
Biomatik Corporation Recombinant Apolipoprotein C3 (APOC3), 100UG
Supplier:  Biomatik Corporation RPU40353
Encompass_Preferred
Recombinant Mouse Apolipoprotein C3 (APOC3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Apolipoprotein C3 (APOC3). Target Synonyms: Apo-C3; APOCIII; APOC-III. Accession Number: P33622. Gene ID: 11814. Expression Region: Glu21~Ser99. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5476