Learn More
Biomatik Corporation Recombinant Apolipoprotein E (APOE), 100UG
Supplier:  Biomatik Corporation RPU40358
Encompass_Preferred
Recombinant Mouse Apolipoprotein E (APOE) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Apolipoprotein E (APOE). Target Synonyms: Apo-E; AD2; Apoprotein; Alzheimer Disease 2(E4-Associated,Late Onset. Accession Number: P08226. Gene ID: 11816. Expression Region: Thr14~Lys167. Tag Info: N-terminal His and GST Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5477