Learn More
Biomatik Corporation Recombinant Aquaporin 1, Colton Blood Group (AQP1), 100UG
Supplier:  Biomatik Corporation RPU40374
Encompass_Preferred
Recombinant Human Aquaporin 1 (AQP1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Aquaporin 1 (AQP1). Target Synonyms: AQP-CHIP; CHIP28; CO; Channel-Forming Integral Protein,28kDa; Aquaporin-CHIP; Urine water channel; Water channel protein for red blood cells and kidney proximal tubule. Accession Number: P29972. Gene ID: 358. Expression Region: Val201~Lys269. Tag Info: N-terminal His and GST Tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5479