Learn More
Biomatik Corporation Recombinant Arginase (ARG), 100UG
Supplier:  Biomatik Corporation RPU40382
Encompass_Preferred
Recombinant Rat Arginase (ARG) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Arginase (ARG). Target Synonyms: ARG1; Arginase I; Liver Arginase. Accession Number: P07824. Gene ID: 29221. Expression Region: Met1~Lys323. Tag Info: N-terminal His Tag. Theoretical MW: 36kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5481