Learn More
Biomatik Corporation Recombinant B-Cell Activating Factor (BAFF), 100UG
Supplier:  Biomatik Corporation RPU40423
Encompass_Preferred
Recombinant Mouse B-Cell Activating Factor (BAFF) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: B-Cell Activating Factor (BAFF). Target Synonyms: CD257; TNFSF13B; BLYS; TALL1; THANK; TNFSF20; ZTNF4; B lymphocyte stimulator; Tumor Necrosis Factor Ligand Superfamily Member 13B; Dendritic cell-derived TNF-like. Accession Number: Q9WU72. Gene ID: 24099. Expression Region: Tyr68~Phe283. Tag Info: N-terminal His Tag. Theoretical MW: 27/22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5483