Learn More
Biomatik Corporation Recombinant Bone Morphogenetic Protein 4 (BMP4), 100UG
Supplier:  Biomatik Corporation RPU40496
Encompass_Preferred
Recombinant Human Bone Morphogenetic Protein 4 (BMP4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Bone Morphogenetic Protein 4 (BMP4). Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Accession Number: P12644. Gene ID: 652. Expression Region: Ser293~Arg408. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5486