Learn More
Biomatik Corporation Recombinant C Reactive Protein (CRP), 100UG
Supplier:  Biomatik Corporation RPU40533
Encompass_Preferred
Recombinant Pig C Reactive Protein (CRP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: C Reactive Protein (CRP). Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Accession Number: O19062. Gene ID: 396842. Expression Region: Gln20~Pro222. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5491