Learn More
Biomatik Corporation Recombinant C4 Binding Protein Alpha (C4BPa), 100UG
Supplier:  Biomatik Corporation RPU40538
Encompass_Preferred
Recombinant Human C4 Binding Protein Alpha (C4BPa) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: C4 Binding Protein Alpha (C4BPa). Target Synonyms: C4BP-a; PRP; C4BP; Complement Component 4 Binding Protein,Alpha; Proline-rich protein. Accession Number: P04003. Gene ID: 722. Expression Region: Gln548~Leu597. Tag Info: N-terminal His Tag. Theoretical MW: 7kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5492