Learn More
Biomatik Corporation Recombinant Carbohydrate Antigen 125 (CA125), 100UG
Supplier:  Biomatik Corporation RPU40592
Encompass_Preferred
Recombinant Mouse Carbohydrate Antigen 125 (CA125) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Carbohydrate Antigen 125 (CA125). Target Synonyms: MUC16; Cancer Antigen 125; Mucin 16,Cell Surface Associated; Ovarian Cancer-Related Tumor Marker CA125; Ovarian Carcinoma Antigen CA125. Accession Number: XM_911929.4. Expression Region: Thr2~Gln258. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5496