Learn More
Biomatik Corporation Recombinant Caspase 9 (CASP9), 100UG
Supplier:  Biomatik Corporation RPU40688
Encompass_Preferred
Recombinant Mouse Caspase 9 (CASP9) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Caspase 9 (CASP9). Target Synonyms: APAF3; ICE-LAP6; MCH6; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 9; Apoptotic protease-activating factor 3; ICE-like apoptotic protease 6. Accession Number: Q4FJK5. Expression Region: Met1~His200. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5502