Learn More
Biomatik Corporation Recombinant Cathelicidin Antimicrobial Peptide (CAMP), 100UG
Supplier:  Biomatik Corporation RPU40707
Encompass_Preferred
Recombinant Human Cathelicidin Antimicrobial Peptide (CAMP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cathelicidin Antimicrobial Peptide (CAMP). Target Synonyms: CAP18; FALL39; HSD26; LL37; 18 kDa Cationic Antimicrobial Protein; Antibacterial protein FALL-39; FALL-39 peptide antibiotic. Accession Number: P49913. Gene ID: 820. Expression Region: Gln31~Ser170. Tag Info: N-terminal His Tag. Theoretical MW: 25/17/14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5503