Learn More
Biomatik Corporation Recombinant Cathelicidin Antimicrobial Peptide (CAMP), 100UG
Catalog No. 501505504
Encompass_Preferred
Recombinant Mouse Cathelicidin Antimicrobial Peptide (CAMP) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cathelicidin Antimicrobial Peptide (CAMP). Target Synonyms: CAP18; FALL39; HSD26; LL37; 18 kDa Cationic Antimicrobial Protein; Antibacterial protein FALL-39; FALL-39 peptide antibiotic. Accession Number: P51437. Gene ID: 12796. Expression Region: Ser31~Glu172. Tag Info: N-terminal His and GST Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5504 Supplier Biomatik Corporation Supplier No. RPU40708
Recombinant Mouse Cathelicidin Antimicrobial Peptide (CAMP) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cathelicidin Antimicrobial Peptide (CAMP). Target Synonyms: CAP18; FALL39; HSD26; LL37; 18 kDa Cationic Antimicrobial Protein; Antibacterial protein FALL-39; FALL-39 peptide antibiotic. Accession Number: P51437. Gene ID: 12796. Expression Region: Ser31~Glu172. Tag Info: N-terminal His and GST Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.