Learn More
Biomatik Corporation Recombinant Cathepsin S (CTSS), 100UG
Supplier:  Biomatik Corporation RPU40737
Encompass_Preferred
Recombinant Mouse Cathepsin S (CTSS) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cathepsin S (CTSS). Target Synonyms: CTS-S. Accession Number: O70370. Gene ID: 13040. Expression Region: Leu123~Glu339. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5505