Learn More
Biomatik Corporation Recombinant Chemerin (CHEM), 100UG
Catalog No. 501505506
Encompass_Preferred
Recombinant Rat Chemerin (CHEM) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Chemerin (CHEM). Target Synonyms: TIG2; RARRES2; Tazarotene-Induced Gene 2; Retinoic Acid Receptor Responder 2,Tazarotene Induced; RAR-responsive protein TIG2. Accession Number: Q5BK77. Gene ID: 297073. Expression Region: Ser25~Gly152. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5506 Supplier Biomatik Corporation Supplier No. RPU40764
Recombinant Rat Chemerin (CHEM) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Chemerin (CHEM). Target Synonyms: TIG2; RARRES2; Tazarotene-Induced Gene 2; Retinoic Acid Receptor Responder 2,Tazarotene Induced; RAR-responsive protein TIG2. Accession Number: Q5BK77. Gene ID: 297073. Expression Region: Ser25~Gly152. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.