Learn More
Biomatik Corporation Recombinant Cholecystokinin (CCK), 100UG
Catalog No. 501505508
Encompass_Preferred
Recombinant Rat Cholecystokinin (CCK) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Cholecystokinin (CCK). Target Synonyms: CCK-PZ. Accession Number: P01355. Gene ID: 25298. Expression Region: Met1~Ser115. Tag Info: N-terminal His and GST Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5508 Supplier Biomatik Corporation Supplier No. RPU40780
Recombinant Rat Cholecystokinin (CCK) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Cholecystokinin (CCK). Target Synonyms: CCK-PZ. Accession Number: P01355. Gene ID: 25298. Expression Region: Met1~Ser115. Tag Info: N-terminal His and GST Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.