Learn More
Biomatik Corporation Recombinant Human Melanoma antigen recognized by T-cells 1 (MLANA), 100UG
Supplier:  Biomatik Corporation RPC20150
Encompass_Preferred
Recombinant Human Melanoma antigen recognized by T-cells 1 (MLANA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MLANA. Target Synonyms: Antigen LB39-AAAntigen SK29-AAProtein Melan-A. Accession Number: Q16655. Expression Region: 1~118aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 27.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4627