Learn More
Biomatik Corporation Recombinant Chromogranin A (CHGA), 100UG
Catalog No. 501505509
Encompass_Preferred
Recombinant Human Chromogranin A (CHGA) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Chromogranin A (CHGA). Target Synonyms: CGA; SP-I; Parathyroid Secretory Protein 1; Vasostatin; Pancreastatin; Parastatin; Pituitary secretory protein I. Accession Number: P10645. Gene ID: 1113. Expression Region: Gln15~Gly148. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5509 Supplier Biomatik Corporation Supplier No. RPU40795
Recombinant Human Chromogranin A (CHGA) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Chromogranin A (CHGA). Target Synonyms: CGA; SP-I; Parathyroid Secretory Protein 1; Vasostatin; Pancreastatin; Parastatin; Pituitary secretory protein I. Accession Number: P10645. Gene ID: 1113. Expression Region: Gln15~Gly148. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.