Learn More
Biomatik Corporation Recombinant Coagulation Factor VIII (F8), 100UG
Catalog No. 501505511
Encompass_Preferred
Recombinant Pig Coagulation Factor VIII (F8) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Coagulation Factor VIII (F8). Target Synonyms: FVIII; AHF; HEMA; F8B; F8C; Antihemophilic Globulin; Hemophilia A; Anti-Hemophilic Factor A; FVIII Procoagulant Component; F8 Protein. Accession Number: P00741. Expression Region: Gly1669~Ala1824. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5511 Supplier Biomatik Corporation Supplier No. RPU40879
Recombinant Pig Coagulation Factor VIII (F8) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Coagulation Factor VIII (F8). Target Synonyms: FVIII; AHF; HEMA; F8B; F8C; Antihemophilic Globulin; Hemophilia A; Anti-Hemophilic Factor A; FVIII Procoagulant Component; F8 Protein. Accession Number: P00741. Expression Region: Gly1669~Ala1824. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.