Learn More
Biomatik Corporation Recombinant Collagen Type I Alpha 1 (COL1a1), 100UG
Catalog No. 501505512
Encompass_Preferred
Recombinant Mouse Collagen Type I Alpha 1 (COL1a1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Collagen Type I Alpha 1 (COL1a1). Target Synonyms: COL-1A1; COL1-A1; COL1A-1; OI4; Collagen Alpha-1(I)chain. Accession Number: P11087. Gene ID: 12842. Expression Region: Lys1225~Val1453. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5512 Supplier Biomatik Corporation Supplier No. RPU40892
Recombinant Mouse Collagen Type I Alpha 1 (COL1a1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Collagen Type I Alpha 1 (COL1a1). Target Synonyms: COL-1A1; COL1-A1; COL1A-1; OI4; Collagen Alpha-1(I)chain. Accession Number: P11087. Gene ID: 12842. Expression Region: Lys1225~Val1453. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.