Learn More
Biomatik Corporation Recombinant Collagen Type X (COL10), 100UG
Catalog No. 501505514
Encompass_Preferred
Recombinant Human Collagen Type X (COL10) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type X (COL10). Target Synonyms: COL10A1; COL10-A1; Schmid Metaphyseal Chondrodysplasia; Collagen Alpha-1(X)chain. Accession Number: Q03692. Gene ID: 1300. Expression Region: Thr547~Met680. Tag Info: N-terminal His Tag. Theoretical MW: 17/34/50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5514 Supplier Biomatik Corporation Supplier No. RPU40912
Recombinant Human Collagen Type X (COL10) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type X (COL10). Target Synonyms: COL10A1; COL10-A1; Schmid Metaphyseal Chondrodysplasia; Collagen Alpha-1(X)chain. Accession Number: Q03692. Gene ID: 1300. Expression Region: Thr547~Met680. Tag Info: N-terminal His Tag. Theoretical MW: 17/34/50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.