Learn More
Biomatik Corporation Recombinant Complement Component 5a (C5a), 100UG
Catalog No. 501505517
Encompass_Preferred
Recombinant Mouse Complement Component 5a (C5a) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Complement Component 5a (C5a). Accession Number: P06684. Gene ID: 15139. Expression Region: Asn679~Arg755. Tag Info: N-terminal His Tag. Theoretical MW: 10kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5517 Supplier Biomatik Corporation Supplier No. RPU40950
Recombinant Mouse Complement Component 5a (C5a) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Complement Component 5a (C5a). Accession Number: P06684. Gene ID: 15139. Expression Region: Asn679~Arg755. Tag Info: N-terminal His Tag. Theoretical MW: 10kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.