Learn More
Biomatik Corporation Recombinant Complement Factor D (CFD), 100UG
Supplier:  Biomatik Corporation RPU40967
Encompass_Preferred
Recombinant Rat Complement Factor D (CFD) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Complement Factor D (CFD). Target Synonyms: CF-D; DF; PFD; ADN; Adipsin; Properdin Factor D; C3 convertase activator. Accession Number: P32038. Gene ID: 54249. Expression Region: Met1~Ala263. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5519