Learn More
Biomatik Corporation Recombinant Complement Factor H (CFH), 100UG
Supplier:  Biomatik Corporation RPU40968
Encompass_Preferred
Recombinant Human Complement Factor H (CFH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Complement Factor H (CFH). Target Synonyms: CF-H; FH; FHL1; ARMD4; ARMS1; CFHL3; HF1; HF2; HUS; H Factor 2; Age-Related Maculopathy Susceptibility 1; Adrenomedullin binding protein. Accession Number: P08603. Gene ID: 3075. Expression Region: Ser860~Arg1231. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5520