Learn More
Biomatik Corporation Recombinant Complement Receptor 1, Erythrocyte (CR1), 100UG
Supplier:  Biomatik Corporation RPU40975
Encompass_Preferred
Recombinant Human Complement Receptor 1, Erythrocyte (CR1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Complement Receptor 1, Erythrocyte (CR1). Target Synonyms: CD35; C3BR; KN; C3b/C4b Receptor,Including Knops Blood Group System; Immune Adherence Receptor. Accession Number: P17927. Gene ID: 1378. Expression Region: Ile1780~Pro2039. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5521