Learn More
Biomatik Corporation Recombinant Connective Tissue Growth Factor (CTGF), 100UG
Supplier:  Biomatik Corporation RPU40985
Encompass_Preferred
Recombinant Human Connective Tissue Growth Factor (CTGF) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Connective Tissue Growth Factor (CTGF). Target Synonyms: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. Accession Number: P29279. Gene ID: 1490. Expression Region: Leu80~Leu200. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5523