Learn More
Biomatik Corporation Recombinant Corticosteroid Binding Globulin (CBG), 100UG
Supplier:  Biomatik Corporation RPU41013
Encompass_Preferred
Recombinant Human Corticosteroid Binding Globulin (CBG) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Corticosteroid Binding Globulin (CBG). Target Synonyms: SERPINA6; Transcortin; Serpin A6; Serpin Peptidase Inhibitor,Clade A,Member 6(Alpha-1 Antiproteinase; Antitrypsin). Accession Number: P08185. Gene ID: 866. Expression Region: Glu218~Arg378. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5526