Learn More
Biomatik Corporation Recombinant C-Type Lectin Domain Family 4, Member C (CLEC4C), 100UG
Supplier:  Biomatik Corporation RPU41046
Encompass_Preferred
Recombinant Human C-Type Lectin Domain Family 4, Member C (CLEC4C) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: C-Type Lectin Domain Family 4, Member C (CLEC4C). Target Synonyms: CD303; CLECSF11,CLECSF7; HECL; DLEC; BDCA2; Blood dendritic cell antigen 2; C-type lectin superfamily member 7; Dendritic lectin. Accession Number: Q8WTT0. Gene ID: 170482. Expression Region: Glu5~Ile152. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5527