Learn More
Biomatik Corporation Recombinant Cystathionine Gamma Lyase (CSE), 100UG
Supplier:  Biomatik Corporation RPU41102
Encompass_Preferred
Recombinant Mouse Cystathionine (CTH) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cystathionine (CTH). Target Synonyms: CSE; Cystathionase; Cysteine-protein sulfhydrase; Gamma-cystathionase. Accession Number: Q8VCN5. Gene ID: 107869. Expression Region: Phe14~Val224. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5528