Learn More
Biomatik Corporation Recombinant Deoxyribonuclease I (DNASE1), 100UG
Supplier:  Biomatik Corporation RPU41169
Encompass_Preferred
Recombinant Human Deoxyribonuclease I (DNASE1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Deoxyribonuclease I (DNASE1). Target Synonyms: DNaseI; DNL1; DRNI; DNase-I; Dornase alfa. Accession Number: P24855. Gene ID: 1773. Expression Region: Gly19~Ala259. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5532