Learn More
Biomatik Corporation Recombinant Desmoglein 3 (DSG3), 100UG
Supplier:  Biomatik Corporation RPU41178
Encompass_Preferred
Recombinant Human Desmoglein 3 (DSG3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Desmoglein 3 (DSG3). Target Synonyms: CDHF6; PVA; 130 kDa pemphigus vulgaris antigen; Cadherin family member 6. Accession Number: P32926. Gene ID: 1830. Expression Region: Glu858~Ile999. Tag Info: N-terminal His and GST Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5534