Learn More
Biomatik Corporation Recombinant Dihydrolipoyl Transacetylase (DLAT), 100UG
Catalog No. 501505536
Encompass_Preferred
Recombinant Mouse Dihydrolipoyl Transacetylase (DLAT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Dihydrolipoyl Transacetylase (DLAT). Target Synonyms: DLTA; PDC-E2; PBC; M2 antigen complex 70 kDa subunit; Dihydrolipoamide acetyltransferase component of pyruvate dehydrogenase complex; Pyruvate dehydrogenase complex E2. Accession Number: Q8BMF4. Gene ID: 235339. Expression Region: Thr368~Trp626. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5536 Supplier Biomatik Corporation Supplier No. RPU41200
Recombinant Mouse Dihydrolipoyl Transacetylase (DLAT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Dihydrolipoyl Transacetylase (DLAT). Target Synonyms: DLTA; PDC-E2; PBC; M2 antigen complex 70 kDa subunit; Dihydrolipoamide acetyltransferase component of pyruvate dehydrogenase complex; Pyruvate dehydrogenase complex E2. Accession Number: Q8BMF4. Gene ID: 235339. Expression Region: Thr368~Trp626. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.