Learn More
Biomatik Corporation Recombinant Dipeptidyl Peptidase IV (DPP4), 100UG
Supplier:  Biomatik Corporation RPU41205
Encompass_Preferred
Recombinant Human Cluster Of Differentiation 26 (CD26) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cluster Of Differentiation 26 (CD26). Target Synonyms: CD26; ADA; DPPIV; DPP-IV; DPP4; ADABP; ADCP2; TP103; Adenosine Deaminase Complexing Protein 2; T-cell activation antigen CD26; Dipeptidyl Peptidase IV. Accession Number: P27487. Gene ID: 1803. Expression Region: Ser484~Val728. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5537