Learn More
Biomatik Corporation Recombinant Elastase 3B (ELA3B), 100UG
Supplier:  Biomatik Corporation RPU41246
Encompass_Preferred
Recombinant Human Elastase 3B (ELA3B) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Elastase 3B (ELA3B). Target Synonyms: CELA3B; Chymotrypsin Like Elastase Family Member 3B; Protease E; Fecal Elastase; Elastase 1; Cholesterol-Binding Pancreatic Protease; Pancreatic Endopeptidase E. Accession Number: P08861. Gene ID: 23436. Expression Region: Val29~His270. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5538