Learn More
Biomatik Corporation Recombinant Human Gamma Actin (ACTG)
Supplier:  Biomatik Corporation RPU40062
Encompass_Preferred
Recombinant Human Gamma Actin (ACTG) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Gamma Actin (ACTG). Target Synonyms: ACT; ACTB; ACTg1; DFNA20; DFNA26; deafness, autosomal dominant 20; deafness, autosomal dominant 26; Actin, cytoplasmic 2. Accession Number: P63261. Gene ID: 71. Expression Region: Met1~Phe375. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9866