Learn More
Biomatik Corporation Recombinant Human Carbonic Anhydrase IX (CA9)
Supplier:  Biomatik Corporation RPU40608
Encompass_Preferred
Recombinant Human Carbonic Anhydrase IX (CA9) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Carbonic Anhydrase IX (CA9). Target Synonyms: CA-IX; CAIX; MN; pMW1; P54/58N; Carbonic Dehydratase; Renal cell carcinoma-associated antigen G250; RCC-Associated Protein G250; Carbonate dehydratase IX. Accession Number: Q16790. Gene ID: 768. Expression Region: Pro59~Asp414. Tag Info: N-terminal His Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9893