Learn More
Biomatik Corporation Recombinant Rabbit Tissue Factor Pathway Inhibitor (TFPI)
Supplier:  Biomatik Corporation RPU44030
Encompass_Preferred
Recombinant Rabbit Tissue Factor Pathway Inhibitor (TFPI) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rabbit (Oryctolagus cuniculus). Target Name: Tissue Factor Pathway Inhibitor (TFPI). Target Synonyms: TFI; EPI; LACI; TFPI1; Lipoprotein-Associated Coagulation Inhibitor; Extrinsic pathway inhibitor. Accession Number: P19761. Gene ID: 100009401. Expression Region: Lys39~Gly275. Tag Info: N-terminal His and GST Tag. Theoretical MW: 58kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0067