Learn More
Biomatik Corporation Recombinant Rat Transmembrane Protein 173 (TMEM173)
Supplier:  Biomatik Corporation RPU44151
Encompass_Preferred
Recombinant Rat Transmembrane Protein 173 (TMEM173) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Transmembrane Protein 173 (TMEM173). Target Synonyms: NET23; ERIS; MPYS; MITA; STING; Endoplasmic Reticulum Interferon Stimulator; Stimulator Of Interferon Genes Protein; Mediator of IRF3 activation. Accession Number: F1M391. Gene ID: 498840. Expression Region: Ala160~Leu363. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0076