Learn More
Biomatik Corporation Recombinant Rat Tumor Necrosis Factor Ligand Superfamily, Member 14 (TNFSF14)
Supplier:  Biomatik Corporation RPU44242
Encompass_Preferred
Recombinant Rat Tumor Necrosis Factor Ligand Superfamily, Member 14 (TNFSF14) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tumor Necrosis Factor Ligand Superfamily, Member 14 (TNFSF14). Target Synonyms: CD258; TR2; HVEML; LIGHT; LTg; Herpes virus entry mediator ligand; Herpesvirus entry mediator ligand. Accession Number: A6KQR4. Gene ID: 301133. Expression Region: Arg58~Ile239. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0079