Learn More
Biomatik Corporation Recombinant Human Cytokine Inducible SH2 Containing Protein (CISH)
Supplier:  Biomatik Corporation RPU50333
Encompass_Preferred
Recombinant Human Cytokine Inducible SH2 Containing Protein (CISH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytokine Inducible SH2 Containing Protein (CISH). Target Synonyms: CIS; CIS-1; G18; SOCS; Suppressor of cytokine signaling. Accession Number: Q9NSE2. Gene ID: 1154. Expression Region: Met1~Leu258. Tag Info: N-terminal His Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0094