Learn More
Biomatik Corporation Recombinant Human S100 Calcium Binding Protein A15 (S100A15)
Supplier:  Biomatik Corporation RPU50389
Encompass_Preferred
Recombinant Human S100 Calcium Binding Protein A15 (S100A15) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: S100 Calcium Binding Protein A15 (S100A15). Target Synonyms: S100A7A; S100A7L1; S100A7f; S100 calcium-binding protein A7-like 1. Accession Number: Q86SG5. Gene ID: 338324. Expression Region: Ser2~Gln101. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0096