Learn More
Biomatik Corporation Recombinant Human Inhibin Alpha (INHa)
Supplier:  Biomatik Corporation RPU51139
Encompass_Preferred
Recombinant Human Inhibin Alpha (INHa) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Inhibin Alpha (INHa). Target Synonyms: INH-A. Accession Number: P05111. Gene ID: 3623. Expression Region: Ala25~Cys363. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0127