Learn More
Biomatik Corporation Human Mannose Binding Lectin (MBL), Active Protein
Supplier:  Biomatik Corporation RPU51460
Encompass_Preferred
Human Mannose Binding Lectin (MBL), Active Protein is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mannose Binding Lectin (MBL). Target Synonyms: MBL2; COLEC1; HSMBPC; MBP1; MBP; Collectin-1; Mannose-binding protein C; Mannan Binding Protein; Mannose-Binding Lectin(Protein C)2,Soluble(Opsonic Defect). Accession Number: P11226. Gene ID: 4153. Expression Region: Leu130~Ile248. Tag Info: N-terminal His and GST Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0142