Learn More
Biomatik Corporation Recombinant Human Dermokine (DMKN)
Supplier:  Biomatik Corporation RPU51922
Encompass_Preferred
Recombinant Human Dermokine (DMKN) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Dermokine (DMKN). Target Synonyms: Epidermis-specific secreted protein SK30/SK89. Accession Number: Q6E0U4. Gene ID: 93099. Expression Region: Gly27~Gly238. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0171