Learn More
Biomatik Corporation Recombinant Mouse Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1)
Supplier:  Biomatik Corporation RPU52845
Encompass_Preferred
Recombinant Mouse Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1). Target Synonyms: TGF-beta1-BP-1; Transforming growth factor beta-1-binding protein 1. Accession Number: Q8CG19. Gene ID: 268977. Expression Region: Asp1415~Glu1712. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0203