Learn More
Biomatik Corporation Recombinant Human Cathepsin B (CTSB)
Supplier:  Biomatik Corporation RPU40715
Encompass_Preferred
Recombinant Human Cathepsin B (CTSB) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cathepsin B (CTSB). Target Synonyms: CTS-B; APPS; CPSB; APP secretase. Accession Number: P07858. Gene ID: 1508. Expression Region: Val129~Asp333. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9897